Learn More
Foxx Life Sciences EZBio Rigid Reservoir for Tangential Flow Filtration (TFF) System, PP, 2.5L Rectangular Welded Carboy, 83B, VersaCap w/ Rigid tubing 1x Mini TC on 3/8"ID x 1/2"OD, 1x Maxi TC on 9/16"IDx3/4"OD, 1x 3/8"HB w/ Bottom Drain Mini TC, Non-Sterile, 1/CS
Catalog No. 5000332743
Encompass_Preferred
The EZBio SUT Rigid Reservoir System is a single-use rigid bioprocess carboy and bottle solution for fluid handling, supporting upstream and downstream processes. It provides a sterile, closed, scalable platform with integrated rigid tubing for secure, kink-free transfer of buffers, media, process fluids.
Available in 1L–115L volumes (PP bottles/carboys), it features VersaCap technology with customizable 2–5 ports (Mini/Maxi TC options) & bottom Tri-Clamp port for efficient drainage. Plug-and-play integration simplifies setup, reduces contamination risk, and speeds workflows.
Features
Made from USP Class VI materials, gamma-sterilizable, autoclavable (15 PSI, 20 min at 121°C)
Rigid dip tube with notch to allow for easy filling and extraction of media
Bottom TC for quick media harvest
Laser marked ports for quick identification
Multi-port cap system
Applications
Bioprocess storage, aseptic transfer
Cell Harvest
Closed collection system
Catalog No. 50-003-32743 Supplier Foxx Life Sciences Supplier No. 5952511FLS
Add to Cart
Add to Cart
The EZBio SUT Rigid Reservoir System is a single-use rigid bioprocess carboy and bottle solution for fluid handling, supporting upstream and downstream processes. It provides a sterile, closed, scalable platform with integrated rigid tubing for secure, kink-free transfer of buffers, media, process fluids.
Available in 1L–115L volumes (PP bottles/carboys), it features VersaCap technology with customizable 2–5 ports (Mini/Maxi TC options) & bottom Tri-Clamp port for efficient drainage. Plug-and-play integration simplifies setup, reduces contamination risk, and speeds workflows.
Features
Made from USP Class VI materials, gamma-sterilizable, autoclavable (15 PSI, 20 min at 121°C)
Rigid dip tube with notch to allow for easy filling and extraction of media
Bottom TC for quick media harvest
Laser marked ports for quick identification
Multi-port cap system
Applications
Bioprocess storage, aseptic transfer
Cell Harvest
Closed collection system