Learn More
Foxx Life Sciences EZBio Vent Filter, 0.22µm Phobic PTFE, Non-Sterile, 50mm, Gamma safe, 5/PK
Supplier:  Foxx Life Sciences 25H1702FLS
Encompass_Preferred
EZBio Vent Filter, 0.22µm Hydrophobic PTFE Non-Sterile, 50mm, Gamma Stable, 5/PK
Catalog No. NC1489558