Learn More
Foxx Life Sciences EZBio Viper Fill Finish Needles Cytiva (Pall) Allegro™ Filling System Replacement, SS316L Canula 1.6mm ID 87.10mm Length, PC Hub, Single Use Filling Needles, Non-Sterile, 50/CS
Catalog No. 5000308418
Encompass_Preferred
EZBio Viper Needles are high-performance bioprocess filling needles compatible with Cytiva (Pall) filling machines and suited for vials, syringes, and bottles. The Single-use filling needles feature an electropolished, passivated 316L stainless steel cannula with a USP Class VI, gamma-stable polycarbonate hub. Made through electropolishing, ultrasonic cleaning, and passivation, they meet strict quality standards.
Features
High-quality fluid dispensing needle for bioprocess fluid handling.
316L stainless steel cannula offers superior precision versus PEEK tubing.
Aseptic filling hose-barb hub ensures a secure, leak-free connection.
Pall filling machine compatible; inner diameters from 0.8–4.8 mm.
Electropolished surface reduces contamination risk.
Cannula bonded to hub with medical-grade UV-curing adhesive for durability.
Packaging: Double Bagged
Applications:
Final fill for drug products
Cell and gene therapy
Formulations
Benchtop filling
Catalog No. 50-003-08418 Supplier Foxx Life Sciences Supplier No. 29C1010FLS
Add to Cart
Add to Cart
Request a Quote
EZBio Viper Needles are high-performance bioprocess filling needles compatible with Cytiva (Pall) filling machines and suited for vials, syringes, and bottles. The Single-use filling needles feature an electropolished, passivated 316L stainless steel cannula with a USP Class VI, gamma-stable polycarbonate hub. Made through electropolishing, ultrasonic cleaning, and passivation, they meet strict quality standards.
Features
High-quality fluid dispensing needle for bioprocess fluid handling.
316L stainless steel cannula offers superior precision versus PEEK tubing.
Aseptic filling hose-barb hub ensures a secure, leak-free connection.
Pall filling machine compatible; inner diameters from 0.8–4.8 mm.
Electropolished surface reduces contamination risk.
Cannula bonded to hub with medical-grade UV-curing adhesive for durability.
Packaging: Double Bagged
Applications:
Final fill for drug products
Cell and gene therapy
Formulations
Benchtop filling