Learn More
Foxx Life Sciences EZBiopure Chemically Resistant Fluoroelastomer (FKM) Supercritical Fluid Transfer Tubing 4.76mm (3/16") ID X 9.5mm (3/8") OD, 25FT
Catalog No. 5000460515
Encompass_Preferred
EZBiopure Fluoroelastomer (FKM) Tubing (popularly known as Viton tubing) is engineered for supercritical fluid transfer and the safe handling of highly corrosive chemicals and solvents in pharmaceutical applications. It is produced in a controlled cleanroom environment to meet stringent food and pharma-grade standards.
Features
Made and packaged in an ISO Class 7 certified cleanroom.
Chemical Resistance: Excellent compatibility with alcohols, acids, and halogenated solvents (e.g., IPA, MDC).
High Safety: Superior flame and fire resistance.
Wide Temperature Range: Operational from –15°C to +250°C.
Applications:
Corrosive chemical transfer
Chlorinated, fluorinated chemical transfer
Chemical and analytical analysis
Fuel and oil industries
Certifications
FDA 21 CFR 177.2600 Certificate
USP Class VI Certificate
Phthalates / Bis Phenol / Plasticizer free Certification
TSE/BSE Free Certification
5 years Shelf Life
Catalog No. 50-004-60515 Supplier Foxx Life Sciences Supplier No. CHE00480095025
EZBiopure Fluoroelastomer (FKM) Tubing (popularly known as Viton tubing) is engineered for supercritical fluid transfer and the safe handling of highly corrosive chemicals and solvents in pharmaceutical applications. It is produced in a controlled cleanroom environment to meet stringent food and pharma-grade standards.
Features
Made and packaged in an ISO Class 7 certified cleanroom.
Chemical Resistance: Excellent compatibility with alcohols, acids, and halogenated solvents (e.g., IPA, MDC).
High Safety: Superior flame and fire resistance.
Wide Temperature Range: Operational from –15°C to +250°C.
Applications:
Corrosive chemical transfer
Chlorinated, fluorinated chemical transfer
Chemical and analytical analysis
Fuel and oil industries
Certifications
FDA 21 CFR 177.2600 Certificate
USP Class VI Certificate
Phthalates / Bis Phenol / Plasticizer free Certification
TSE/BSE Free Certification
5 years Shelf Life