Learn More
Sigma Aldrich Fine Chemicals Biosciences EZBlueTM Gel Staining Reag
Catalog No. 502904827
Encompass
Convenient sensitive and safe EZBlue(TM) Coomassie Brilliant Blue G-250 colloidal protein stain improves protein electrophoresis results while significantly reducing staining time. Conveniently packaged EZBlue requires no messy weigh-ups or additions of methanol or acid. As a colloidal stain it reacts only with proteins not the gel itself. Background staining is reduced so protein bands can be visualized almost immediately. No destaining step is required although a water wash may intensify bands and clarify the background. Most impressively EZBlue is extremely sensitive detecting as little as 5 ng of protein.
Catalog No. 50-290-4827 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. G10413.8L
Convenient sensitive and safe EZBlue(TM) Coomassie Brilliant Blue G-250 colloidal protein stain improves protein electrophoresis results while significantly reducing staining time. Conveniently packaged EZBlue requires no messy weigh-ups or additions of methanol or acid. As a colloidal stain it reacts only with proteins not the gel itself. Background staining is reduced so protein bands can be visualized almost immediately. No destaining step is required although a water wash may intensify bands and clarify the background. Most impressively EZBlue is extremely sensitive detecting as little as 5 ng of protein.