Learn More
Foxx Life Sciences EZCellGene Single Use (SUT) Media Bottle Assembly, 500mL, PC, 38-430 VersaCap with Overmolded Platinum Cured Silicone Small Bore Tubing 1/16" ID x 3/16" OD & Luer, Sterile, 10/CS
GREENER_CHOICE
Catalog No. 5000388293
Encompass_Preferred
The EZCellGene 38-430 Media Bottle Assembly is a ready-to-use, single-use solution for media & reagent transfer in cell culture & bioprocessing workflows. Each assembly features VersaCap technology with two integrated 1/16" ID hose barbs for vent filtration & fluid transfer. Assemblies are available with Plug, male or female Luer lock, or MicroCNX Connector by CPC to support flexible process connections. All components are made from USP Class VI–compliant materials & are gamma irradiated for immediate use.
Features
Compatible with 38-430 neck bottles for secure, leak-free connections
Over-molded silicone interface maintains a continuous sterile fluid path
Small-bore tubing (1/16" ID × 3/16" OD) enables precise, low-dead-volume flow
Low-shear fluid transfer suitable for sensitive cells, including stem cells
Applications
Cell culture
Feeding, aseptic sampling & fluid transfer
Cell & gene therapy
Small-volume bioreactors & media bottle setups
Catalog No. 50-003-88293 Supplier Foxx Life Sciences Supplier No. 6386002FLS
The EZCellGene 38-430 Media Bottle Assembly is a ready-to-use, single-use solution for media & reagent transfer in cell culture & bioprocessing workflows. Each assembly features VersaCap technology with two integrated 1/16" ID hose barbs for vent filtration & fluid transfer. Assemblies are available with Plug, male or female Luer lock, or MicroCNX Connector by CPC to support flexible process connections. All components are made from USP Class VI–compliant materials & are gamma irradiated for immediate use.
Features
Compatible with 38-430 neck bottles for secure, leak-free connections
Over-molded silicone interface maintains a continuous sterile fluid path
Small-bore tubing (1/16" ID × 3/16" OD) enables precise, low-dead-volume flow
Low-shear fluid transfer suitable for sensitive cells, including stem cells
Applications
Cell culture
Feeding, aseptic sampling & fluid transfer
Cell & gene therapy
Small-volume bioreactors & media bottle setups