Learn More
Foxx Life Sciences EZCellGene TR Single Use (SUT) Media Bottle Assembly, 1L, PS, GL45 wide mouth VersaCap with Overmolded Platinum Cured Silicone Small Bore Tubing 1/16" ID x 3/16" OD & MicroCNX, Sterile, 10/CS
GREENER_CHOICE
Catalog No. 5000388302
Encompass_Preferred
The EZCellGene GL45 wide-mouth media Bottle Assembly is a single-use, preconfigured solution designed to simplify media & reagent delivery in bioprocess & cell culture workflow. Each assembly features VersaCap technology with two integrated 1/16" ID hose barbs for vent filtration & fluid transfer. Assemblies are available with Plug, male/female Luer lock, or MicroCNX Connector by CPC for process connections. All components are made from USP Class VI–compliant materials & gamma irradiated for immediate use.
Features
Designed for GL45 wide-mouth neck bottles for secure & leak-free connections
Over-molded silicone interface maintains a continuous sterile fluid path
Small-bore tubing (1/16"ID × 3/16"OD) enables precise, low-dead-volume flow
Low-shear fluid transfer suitable for sensitive cells, including stem cells
Applications
Cell culture
Feeding, aseptic sampling & fluid transfer
Cell & gene therapy
Small-volume bioreactors & media bottle setups
Catalog No. 50-003-88302 Supplier Foxx Life Sciences Supplier No. 6367304FLS
The EZCellGene GL45 wide-mouth media Bottle Assembly is a single-use, preconfigured solution designed to simplify media & reagent delivery in bioprocess & cell culture workflow. Each assembly features VersaCap technology with two integrated 1/16" ID hose barbs for vent filtration & fluid transfer. Assemblies are available with Plug, male/female Luer lock, or MicroCNX Connector by CPC for process connections. All components are made from USP Class VI–compliant materials & gamma irradiated for immediate use.
Features
Designed for GL45 wide-mouth neck bottles for secure & leak-free connections
Over-molded silicone interface maintains a continuous sterile fluid path
Small-bore tubing (1/16"ID × 3/16"OD) enables precise, low-dead-volume flow
Low-shear fluid transfer suitable for sensitive cells, including stem cells
Applications
Cell culture
Feeding, aseptic sampling & fluid transfer
Cell & gene therapy
Small-volume bioreactors & media bottle setups