Learn More
Bellco Glass, Inc. EZclear 10L (2.5 Gal) BPA-Free PETG Space Saving Carboy with 83mm (83B) VersaCap and 13L Max Capacity with Spigot Large Pinched Handles,1/EA
SDP
Supplier:  Bellco Glass, Inc. 452800343
Encompass_Preferred
The EZgrip Carboys are designed to provide users with a highly versatile container that maximizes storage efficiency and ease of use. The distinctive rectangular shape saves valuable bench space. Larger handles on top of the container and molded grips in the base provide a full, secure grip to make lifting, handling, and pouring easy. The optional wide neck allows the user to reach inside the carboy for easier cleaning. Carboy material and volume are clearly imprinted on the outside of the bottle as well as large, easy to read, metric graduation marks which are certified to ±5% accuracy.PETG carboys are made from a USP Class VI, FDA Grade, BPA-Free materials. Height (w/Cap): 19.7" (501mm)Height (w/o Cap) 19.4" (492mm)Length: 10.75" (273mm)Width: 6.38" (162mm)Approximate Capacity(max): 13LCarboy Material: PETGCap Material: PolypropyleneSpigot Material: Polypropylene
Catalog No. 50-255-0419