Learn More
Bellco Glass, Inc. EZclear 5L (1 Gal) BPA-Free PETG Space Saving Carboy with 83mm (83B) VersaCap and 6.9L Max Capacity, with Spigot and Large Pinched Handle, 1/EA
SDP
Supplier:  Bellco Glass, Inc. 452800341
Encompass_Preferred
The EZgrip Carboys are designed to provide users with a highly versatile container that maximizes storage efficiency and ease of use. The distinctive rectangular shape saves valuable bench space. Larger handles on top of the container and molded grips in the base provide a full, secure grip to make lifting, handling, and pouring easy. The optional wide neck allows the user to reach inside the carboy for easier cleaning. Carboy material and volume are clearly imprinted on the outside of the bottle as well as large, easy to read, metric graduation marks which are certified to ±5% accuracy.PETG carboys are made from a USP Class VI, FDA Grade, BPA-Free materials. Height (w/Cap): 16.4" (417mm)Height (w/o Cap) 16.1" (406mm)Length: 8.66" (220mm)Width: 5.16" (131mm)Approximate Capacity(max): 6.9LCarboy Material: PETGCap Material: Polypropylene
Catalog No. 50-255-0421