Learn More
Apexbio Technology LLC Ezetimibe 163222-33-1 10mM (in 1mL DMSO)
SDP
Catalog No. 5000061779
Encompass_Preferred
Ezetimibe (CAS 163222-33-1) is a cholesterol absorption inhibitor that targets the intestinal cholesterol transporter Niemann-Pick C1 Like 1 (NPC1L1) It reduces intestinal uptake of cholesterol and dietary carotenoids including - and -carotene lycopene lutein and -cryptoxanthin through downregulating transport proteins such as SR-BI ABCA1 NPC1L1 and transcription factors including SREBP-1/-2 and LXR- In ApoE-deficient mouse models ezetimibe significantly decreases plasma cholesterol levels and attenuates atherosclerotic lesion formation Clinically it lowers LDL cholesterol total cholesterol and triglycerides while elevating HDL cholesterol
Catalog No. 50-000-61779 Supplier Apexbio Technology LLC Supplier No. A84305.1
Ezetimibe (CAS 163222-33-1) is a cholesterol absorption inhibitor that targets the intestinal cholesterol transporter Niemann-Pick C1 Like 1 (NPC1L1) It reduces intestinal uptake of cholesterol and dietary carotenoids including - and -carotene lycopene lutein and -cryptoxanthin through downregulating transport proteins such as SR-BI ABCA1 NPC1L1 and transcription factors including SREBP-1/-2 and LXR- In ApoE-deficient mouse models ezetimibe significantly decreases plasma cholesterol levels and attenuates atherosclerotic lesion formation Clinically it lowers LDL cholesterol total cholesterol and triglycerides while elevating HDL cholesterol