Learn More
Medchemexpress LLC Ezetimibe-d4-1 | 1093659-89-2 | MFCD08063570 | 99.3% | 413.45 | 5 MG
SDP
Catalog No. 5000384648
Encompass_Preferred
Ezetimibe-d4-1 is deuterium labeled Ezetimibe. Ezetimibe (SCH 58235) is a potent cholesterol absorption inhibitor, a Niemann-Pick C1-like1 (NPC1L1) inhibitor, and a potent Nrf2 activator. This product is for research use only and not for sale to patients.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration has the potential to affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-003-84648 Supplier Medchemexpress LLC Supplier No. HY17376S15MG
Ezetimibe-d4-1 is deuterium labeled Ezetimibe. Ezetimibe (SCH 58235) is a potent cholesterol absorption inhibitor, a Niemann-Pick C1-like1 (NPC1L1) inhibitor, and a potent Nrf2 activator. This product is for research use only and not for sale to patients.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration has the potential to affect the pharmacokinetic and metabolic profiles of drugs.