Learn More
Medchemexpress LLC Ezetimibe-d4-1 | 1093659-89-2 | MFCD08063570 | 99.3% | 413.45 | C24H17D4F2NO3 | 10 MG
SDP
Catalog No. 5000299174
Encompass_Preferred
Ezetimibe-d4-1 is a deuterium-labeled analogue of ezetimibe used as an analytical internal standard for quantitative mass spectrometry and method development. It provides a defined mass shift for accurate quantitation and is supplied with high reported purity suitable for bioanalytical applications.
  • Deuterium-labeled internal standard for LC-MS/MS.
  • High purity (99.3%) for reliable analytical results.
  • Molecular weight 413.45 for precise mass calibration.
  • Powder formulation with storage recommended at -20°C for long-term stability.
  • Suitable for method validation and pharmacokinetic studies.
  • Available in small pack sizes for laboratory use.
Catalog No. 50-002-99174 Supplier Medchemexpress LLC Supplier No. HY17376S110MG
Ezetimibe-d4-1 is a deuterium-labeled analogue of ezetimibe used as an analytical internal standard for quantitative mass spectrometry and method development. It provides a defined mass shift for accurate quantitation and is supplied with high reported purity suitable for bioanalytical applications.
  • Deuterium-labeled internal standard for LC-MS/MS.
  • High purity (99.3%) for reliable analytical results.
  • Molecular weight 413.45 for precise mass calibration.
  • Powder formulation with storage recommended at -20°C for long-term stability.
  • Suitable for method validation and pharmacokinetic studies.
  • Available in small pack sizes for laboratory use.