Learn More
Medchemexpress LLC Ezetimibe-d4-1 (SCH 58235-d4-1) | 1093659-89-2 | 99.3% | 413.45 | 1 MG
SDP
Catalog No. 5000384232
Encompass_Preferred
Ezetimibe-d4-1 is deuterium labeled Ezetimibe, a potent cholesterol absorption inhibitor and a Niemann-Pick C1-like1 (NPC1L1) inhibitor. It also acts as a potent Nrf2 activator. This compound is for research use only and not sold to patients.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-84232 Supplier Medchemexpress LLC Supplier No. HY17376S11MG
Ezetimibe-d4-1 is deuterium labeled Ezetimibe, a potent cholesterol absorption inhibitor and a Niemann-Pick C1-like1 (NPC1L1) inhibitor. It also acts as a potent Nrf2 activator. This compound is for research use only and not sold to patients.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS