Learn More
Cayman Chemical Ezetimibe-d4, Purity: ≥99% deuterated forms (d1-d4), CAS: 1093659-89-2, 1 mg
SDP
Catalog No. 502933749
Encompass_Preferred
An internal standard for the quantification of ezetimibe by GC- or LC-MS
Catalog No. 50-293-3749 Supplier Cayman Chemical Supplier No. 250441 MG
An internal standard for the quantification of ezetimibe by GC- or LC-MS