Learn More
Medchemexpress LLC Ezetimibe-d4 (SCH 58235-d4) | 1093659-90-5 | 98.84% | 413.45 | 1 MG
SDP
Catalog No. 5000384719
Encompass_Preferred
Ezetimibe-d4 (SCH 58235-d4) is the deuterium labeled Ezetimibe (HY-17376). Ezetimibe (SCH 58235) is a potent cholesterol absorption inhibitor, a Niemann-Pick C1-like1 (NPC1L1) inhibitor, and a potent Nrf2 activator.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements incorporated into drug molecules for quantitation during drug development.
  • Deuteration has potential to affect pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-003-84719 Supplier Medchemexpress LLC Supplier No. HY17376S1MG
Ezetimibe-d4 (SCH 58235-d4) is the deuterium labeled Ezetimibe (HY-17376). Ezetimibe (SCH 58235) is a potent cholesterol absorption inhibitor, a Niemann-Pick C1-like1 (NPC1L1) inhibitor, and a potent Nrf2 activator.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements incorporated into drug molecules for quantitation during drug development.
  • Deuteration has potential to affect pharmacokinetic and metabolic profiles of drugs.