Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ezetimibe ketone | 191330-56-0 | MFCD13185925 | 100.0% | 407.41 g/mol | C24H19F2NO3 | 5 MG
SDP

Catalog No. 5000262066
Encompass_Preferred

Ezetimibe ketone is a phase-I metabolite of ezetimibe that functions as an inhibitor of NPC1L1 and is used in research to study cholesterol absorption and metabolism. It is provided as a high-purity reference material for in vitro and analytical applications and is not for human use.

  • Phase-I metabolite of ezetimibe.
  • NPC1L1 inhibitor that reduces intestinal cholesterol absorption.
  • Reported purity 99.97%.
  • Molecular formula C24H19F2NO3.
  • Molecular weight 407.41 g/mol.
  • Supplied in small research quantities, such as 5 mg.

Catalog No. 50-002-62066 Supplier Medchemexpress LLC Supplier No. HY1331145MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ezetimibe ketone is a phase-I metabolite of ezetimibe that functions as an inhibitor of NPC1L1 and is used in research to study cholesterol absorption and metabolism. It is provided as a high-purity reference material for in vitro and analytical applications and is not for human use.

  • Phase-I metabolite of ezetimibe.
  • NPC1L1 inhibitor that reduces intestinal cholesterol absorption.
  • Reported purity 99.97%.
  • Molecular formula C24H19F2NO3.
  • Molecular weight 407.41 g/mol.
  • Supplied in small research quantities, such as 5 mg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.