Learn More
Medchemexpress LLC Ezetimibe ketone | 191330-56-0 | MFCD13185925 | 100.0% | C24H19F2NO3 | 10MG
SDP
Catalog No. 5000259688
Encompass_Preferred
Ezetimibe ketone is a phase-I metabolite of ezetimibe used as a research reagent. It acts as an NPC1L1 inhibitor and Nrf2 activator, and is employed in studies of cholesterol absorption, drug metabolism, and cellular signaling. Supplied as a high-purity solid and as a DMSO solution, it is intended for in vitro research applications.
  • High purity suitable for analytical and biological assays.
  • Available as solid and as a DMSO solution for assay flexibility.
  • Applicable as a reference standard and metabolic intermediate in research.
  • Supports studies of cholesterol transport, metabolism, and signaling pathways.
  • Characterized with a CAS registry number for unambiguous identification.
Catalog No. 50-002-59688 Supplier Medchemexpress LLC Supplier No. HY13311410MG
Ezetimibe ketone is a phase-I metabolite of ezetimibe used as a research reagent. It acts as an NPC1L1 inhibitor and Nrf2 activator, and is employed in studies of cholesterol absorption, drug metabolism, and cellular signaling. Supplied as a high-purity solid and as a DMSO solution, it is intended for in vitro research applications.
  • High purity suitable for analytical and biological assays.
  • Available as solid and as a DMSO solution for assay flexibility.
  • Applicable as a reference standard and metabolic intermediate in research.
  • Supports studies of cholesterol transport, metabolism, and signaling pathways.
  • Characterized with a CAS registry number for unambiguous identification.