Learn More
Medchemexpress LLC HY-135391 1mg , Ezetimibe phenoxy glucuronide CAS:190448-57-8 Purity:>98%
SDP
Catalog No. 502261782
Encompass_Preferred
HY-135391 1mg Ezetimibe phenoxy glucuronide CAS: 190448-57-8 Ezetimibe phenoxy glucuronide (Ezetimibe glucuronide) is the active metabolite of Ezetimibe. Antihyperlipoproteinemic activity. Ezetimibe is a potent cholesterol absorption inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
This item is not returnable. View return policy
Catalog No. 50-226-1782 Supplier Medchemexpress LLC Supplier No. HY1353911MG
HY-135391 1mg Ezetimibe phenoxy glucuronide CAS: 190448-57-8 Ezetimibe phenoxy glucuronide (Ezetimibe glucuronide) is the active metabolite of Ezetimibe. Antihyperlipoproteinemic activity. Ezetimibe is a potent cholesterol absorption inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.