Learn More
Foxx Life Sciences Foxx Life Sciences™ Carboy Vent Filters
Catalog No. NC2020205
Encompass_Preferred
Foxx Life Sciences™ Carboy Vent Filters are used for hydrophobic venting and solvent filtration applications. The PTFE filter membrane is ideal for maintaining purified water stored in carboys, or for sterile venting during a slow exhaust/liquid autoclave cycle of EZgrip carboys.
Catalog No. NC2020205 Supplier Foxx Life Sciences Supplier No. 25H1628OEM
Foxx Life Sciences™ Carboy Vent Filters are used for hydrophobic venting and solvent filtration applications. The PTFE filter membrane is ideal for maintaining purified water stored in carboys, or for sterile venting during a slow exhaust/liquid autoclave cycle of EZgrip carboys.