Learn More
Bellco Glass, Inc. EZFlow Syringe Filter, 0.45µm Glass Fiber, 25mm, 100/PK
SDP
Supplier:  Bellco Glass, Inc. 452800550
Encompass_Preferred
EZFlow 13mm, 25mm, and 33mm Syringe Filters are used for prefiltration, clarification, or fine particulate removal. These syringe filters are available in a variety of filter membrane materials, offered in 0.2µm or 0.45µm pore sizes. The filter membranes are reinforced with polypropylene housing with a female Luer-lok inlet and a male Luer slip outlet.All materials comply with USP Class VI Standards and are available for both sterile and non-sterile applications.USP Class VISpecificationsProduct Diameter: 1.17" (29.7mm)Product Height: 0.88" (22.4mm)Filter Diameter: 25mmFilter Material: Glass FiberFilter Pore Size: .45µm
Catalog No. 50-255-0212