Learn More
Bellco Glass, Inc. EZFlow Syringe Filter-Sample Prep, 0.45µm Hydrophilic PTFE, 33mm, 100/PK
SDP
Supplier:  Bellco Glass, Inc. 452800570
Encompass_Preferred
EZFlow 13mm, 25mm, and 33mm sample prep syringe filters are designed for aqueous and organic solution samples. The sample prep syringe filters are used for clarification and particle removal from samples before instrumental analysis such as HPLC, IC, and GC. These syringe filters are available in a variety of filter membrane materials, offered in 0.2µm or 0.45µm pore sizes. The filter membranes are reinforced with polypropylene housing with a female Luer-lok inlet and a male Luer slip outlet.All materials comply with USP Class VI Standards.USP Class VISpecificationsProduct Diameter: 1.31" (33mm)Product Height: 1.02" (26mm)Filter Diameter: 33mmFilter Material: Hydrophilic PTFEFilter Pore Size: .45µm
Catalog No. 50-255-0192