Learn More
Foxx Life Sciences EZGrip 115L (30 Gal) Carboy, HDPE, Wide Mouth 120mm VersaCap, 120L Max Capacity, Space Saving Rectangular Carboy with Handles and Graduation Marks, Non Sterile, 1/EA
Catalog No. 5000078001
Encompass_Preferred
Foxx Life Sciences EZGrip Carboy is a versatile fluid handling system designed for efficiency and ease of use. Made from USP Class VI, FDA-grade materials, it’s available sterile or non-sterile.
The rectangular shape saves bench space, and the wide-mouth design allows easy filling and cleaning. Its ergonomic body includes top handles and molded grips for safe handling. Each carboy features VersaCap Technology with leak-proof, interchangeable top connections and adapter inserts. Graduations are certified to ±5% accuracy.Designed for bioprocessing and fluid storage. Also functions as a lab fluid handling container or a laboratory carboy
Features
Gamma sterilized, cleanroom double-bagged (sterile models)
Ergonomic design with molded grips
Wide-mouth for easy use
Certified graduation marks
Applications
Bioprocess container for media formulation and mixing
Aseptic processes
Handling aggressive chemicals
Production with low leachables/extractables
Catalog No. 50-000-78001 Supplier Foxx Life Sciences Supplier No. 1528121FLS
Foxx Life Sciences EZGrip Carboy is a versatile fluid handling system designed for efficiency and ease of use. Made from USP Class VI, FDA-grade materials, it’s available sterile or non-sterile.
The rectangular shape saves bench space, and the wide-mouth design allows easy filling and cleaning. Its ergonomic body includes top handles and molded grips for safe handling. Each carboy features VersaCap Technology with leak-proof, interchangeable top connections and adapter inserts. Graduations are certified to ±5% accuracy.Designed for bioprocessing and fluid storage. Also functions as a lab fluid handling container or a laboratory carboy
Features
Gamma sterilized, cleanroom double-bagged (sterile models)
Ergonomic design with molded grips
Wide-mouth for easy use
Certified graduation marks
Applications
Bioprocess container for media formulation and mixing
Aseptic processes
Handling aggressive chemicals
Production with low leachables/extractables