Learn More
Foxx Life Sciences EZLabpure 50L Round Carboy, PP, VersaCap 83B, Wide Mouth Carboy with handle and graduation marks, Mixing Carboy, Autoclavable, Non Sterile, 1EA
GREENER_CHOICE
Catalog No. 502551737
Encompass_Preferred
Foxx Life Sciences offers a premium line of high quality, Round Polypropylene (PP) laboratory Carboys comes with patented VersaCap technology. VersaCap offers large handles with finger grips that allow for easy tightening and opening. Our versatile leak proof carboys with strong easy-grip handles, aim to take full advantage of storage efficiency at minimal effort. Overall functionality is available for many applications, in 10 Liter or 20 Liter sizes, with or without spigots each with a VersaCap 83B Polypropylene (PP) Cap. 20 Liter Carboy comes with Graduation marks.
Features
Manufactured using high purity virgin USP Class VI Medical grade polypropylene
These carboys are used for storing and shipping reagents, chemicals, culture media, and standards
PP Carboys are autoclavable at 121°C, at 15 psi for 20 mins.
Can be used at a temperature below 0° C
Superior chemical resistance
Catalog No. 50-255-1737 Supplier Foxx Life Sciences Supplier No. 155E191FLS
Foxx Life Sciences offers a premium line of high quality, Round Polypropylene (PP) laboratory Carboys comes with patented VersaCap technology. VersaCap offers large handles with finger grips that allow for easy tightening and opening. Our versatile leak proof carboys with strong easy-grip handles, aim to take full advantage of storage efficiency at minimal effort. Overall functionality is available for many applications, in 10 Liter or 20 Liter sizes, with or without spigots each with a VersaCap 83B Polypropylene (PP) Cap. 20 Liter Carboy comes with Graduation marks.
Features
Manufactured using high purity virgin USP Class VI Medical grade polypropylene
These carboys are used for storing and shipping reagents, chemicals, culture media, and standards
PP Carboys are autoclavable at 121°C, at 15 psi for 20 mins.
Can be used at a temperature below 0° C
Superior chemical resistance