Learn More
Foxx Life Sciences EZLabpure™ Peroxide Cured Silicone Transparent Tubing, 3mm (1/8") ID X 6mm (1/4") OD, 50FT
Catalog No. 5000468780
Encompass_Preferred
EZLabpure™ Peroxide-Cured Silicone Tubing is designed for general, non-critical fluid transfer applications. This translucent tubing offers excellent flexibility and resiliency, making it ideal for peristaltic pump use in routine non critical processes.
Features
Made from high-quality peroxide-cured silicone rubber.
Manufactured and packaged in a dust-free facility.
Odorless and tasteless for clean fluid transfer.
Exhibits good compression set, tensile strength, and flex fatigue resistance to extend pump life.
Wide Temperature Range: Operational from –80°C to +230°C.
Applications:
Peristaltic Pump
Liquid filling and transfer in Pharma & Biopharma Industries
Food and Beverages Industries
Certifications
Phthalates / Bis Phenol / Plasticizer free Certification
TSE/BSE Free Certification
5 years Shelf Life
FDA 21 CFR 177.2600 Certificate
Catalog No. 50-004-68780 Supplier Foxx Life Sciences Supplier No. PEX00300060050
EZLabpure™ Peroxide-Cured Silicone Tubing is designed for general, non-critical fluid transfer applications. This translucent tubing offers excellent flexibility and resiliency, making it ideal for peristaltic pump use in routine non critical processes.
Features
Made from high-quality peroxide-cured silicone rubber.
Manufactured and packaged in a dust-free facility.
Odorless and tasteless for clean fluid transfer.
Exhibits good compression set, tensile strength, and flex fatigue resistance to extend pump life.
Wide Temperature Range: Operational from –80°C to +230°C.
Applications:
Peristaltic Pump
Liquid filling and transfer in Pharma & Biopharma Industries
Food and Beverages Industries
Certifications
Phthalates / Bis Phenol / Plasticizer free Certification
TSE/BSE Free Certification
5 years Shelf Life
FDA 21 CFR 177.2600 Certificate