Learn More
Foxx Life Sciences EZLabpure™ Platinum-Cured Silicone Tubing, 9mm (23/64") ID X 16mm (5/8") OD, 100FT
Supplier: Foxx Life Sciences FLO00900160100
EZLabpure™ Platinum-Cured Silicone Tubing is designed for general-purpose fluid transfer in food & pharmaceutical applications. Its smooth inner bore minimizes particle entrapment compared to peroxide-cured tubing, ensuring cleaner, more efficient flow. The tubing is developed to meet non-regulatory market requirements while maintaining high performance & reliability.
Features
Made from general-purpose platinum-grade silicone rubber.
Made & packaged in an ISO Class 7 certified cleanroom.
Smoother bore provides superior fluid transfer versus peroxide-cured tubing.
Offers better transparency & longer shelf life than peroxide tubing.
Free from heavy metals & hazardous substances.
Applications:
Peristaltic pump-based fluid transfer
Chemical & analytical analysis
Sanitary application
Food & beverages transfer
Certifications
NSF-51 Certification on Resin
FDA 21 CFR 177.2600
Phthalate/Bisphenol Free COA
TSE/BSE Certification