Learn More
Apexbio Technology LLC EZLink NHS-Biotin Kit, 10 reactions. Cas: MFCD: NA. Amine-reactive biotinyltation kit
SDP
Catalog No. 501905527
Encompass_Preferred
The EZLink NHS-Biotin Kit contains reagents sufficient for 10 biotin labeling reactions. It is a water-soluble biotin reagent for labeling protein, antibody or other molecules with amidogen (NH2-). The biotin in the kit is sufficient and has been activated for direct use. The whole procedure is simple and quick. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5527 Supplier Apexbio Technology LLC Supplier No. K100210
The EZLink NHS-Biotin Kit contains reagents sufficient for 10 biotin labeling reactions. It is a water-soluble biotin reagent for labeling protein, antibody or other molecules with amidogen (NH2-). The biotin in the kit is sufficient and has been activated for direct use. The whole procedure is simple and quick. Other sizes are available. Please inqury us for quote.