Learn More
Medchemexpress LLC HY-111109 5mg Medchemexpress, EZM 2302 CAS:1628830-21-6 Purity:>98%
SDP
Catalog No. 501968823
Encompass_Preferred
Medchemexpress, HY-111109 5mg EZM 2302 CAS:1628830-21-6 EZM 2302 is an inhibitor of coactivator-associated arginine methyltransferase 1 (CARM1) with an IC50 of 6 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8823 Supplier Medchemexpress LLC Supplier No. HY1111095MG
Medchemexpress, HY-111109 5mg EZM 2302 CAS:1628830-21-6 EZM 2302 is an inhibitor of coactivator-associated arginine methyltransferase 1 (CARM1) with an IC50 of 6 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.