Learn More
Medchemexpress LLC HY-144858 5mg , EZM0414 CAS:2411748-50-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1448585MG
Encompass_Preferred
HY-144858 5mg EZM0414 CAS: 2411748-50-8 EZM0414 is a potent, selective, orally bioavailable inhibitor of SETD2 (IC50=18 nM in SETD2 biochemical assay; IC50=34 nM in cellular assay). EZM0414 can be used for the research of relapsed or refractory multiple myeloma and diffuse large B-cell lymphoma. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1853