Learn More
Medchemexpress LLC EZM0414 TFA | 2411759-92-5 | >97.2% | 514.51 | 1 ML
SDP
Catalog No. 5000348208
Encompass_Preferred
EZM0414 TFA, also known as SETD2-IN-1 TFA, is a potent, selective, and orally active inhibitor of SETD2 (IC50=18 nM in biochemical assay; IC50=34 nM in cellular assay). It is used for research into relapsed or refractory multiple myeloma and diffuse large B-cell lymphoma.
  • Potent, selective, and orally active SETD2 inhibitor.
  • Inhibits proliferation of MM and DLBCL cell lines.
  • Shows robust tumor growth regressions in xenograft models.
  • Well-tolerated in animal studies.
  • Exhibits high oral bioavailability in rats and mice.
  • Soluble in DMSO and water for in vitro applications.
  • Provides various in vivo solubility protocols for clear solutions.
Catalog No. 50-003-48208 Supplier Medchemexpress LLC Supplier No. HY13632810MM/1ML
EZM0414 TFA, also known as SETD2-IN-1 TFA, is a potent, selective, and orally active inhibitor of SETD2 (IC50=18 nM in biochemical assay; IC50=34 nM in cellular assay). It is used for research into relapsed or refractory multiple myeloma and diffuse large B-cell lymphoma.
  • Potent, selective, and orally active SETD2 inhibitor.
  • Inhibits proliferation of MM and DLBCL cell lines.
  • Shows robust tumor growth regressions in xenograft models.
  • Well-tolerated in animal studies.
  • Exhibits high oral bioavailability in rats and mice.
  • Soluble in DMSO and water for in vitro applications.
  • Provides various in vivo solubility protocols for clear solutions.