Learn More
Medchemexpress LLC HY-136328 5mg , EZM0414 (TFA) CAS:2411759-92-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1363285MG
Encompass_Preferred
HY-136328 5mg EZM0414 (TFA) CAS: 2411759-92-5 EZM0414 TFA is a potent, selective and orally active inhibitor of SETD2 which is a human histone methyltransferase. EZM0414 TFA has anti-proliferative effects. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9032