Learn More
Omega Biotek Inc Endo-free Plasmid DNA Mini Kit II 50 PREPS
Catalog No. NC1825206
Encompass
The E.Z.N.A. Endo-Free Plasmid Mini Kit II integrates an efficient endotoxin removal step into the plasmid purification procedure to produce high-quality transfection grade (<1 EU/µg) plasmid for efficient transfection.
Catalog No. NC1825206 Supplier Omega Biotek Inc Supplier No. D695001
The E.Z.N.A. Endo-Free Plasmid Mini Kit II integrates an efficient endotoxin removal step into the plasmid purification procedure to produce high-quality transfection grade (<1 EU/µg) plasmid for efficient transfection.