Learn More
MilliporeSigma™ Supelco™ EZSafe™ Flash-Chromatography Assembly, Fritted Disc (Porosity C)
Catalog No. 111007549 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Capacity (Metric):
1000 mL
2000 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
11-100-7549 2000 mL
11-100-7686 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-100-7549 Supplier MilliporeSigma™ Supelco™ Supplier No. Z4164361EA
Add to Cart
Add to Cart
Flash chromatography is useful for rapid, preparative separations with moderate resolutions.
EZSafe™ columns are improved low-pressure columns (max. pressure 7 psig) with unbreakable PTFE fittings and screw-thread connections for increased safety and economy. Bottom tip breakage is eliminated because the valve and exposed drip-tip is all PTFE. GL-45 screw-thread connections ensure positive reliable connections. Eliminates frozen PTFE and glass plugs. Eliminates grease. Choose between standard columns or columns with a fritted disc. The assemblies are supplied with bottom PTFE valve assembly, GL-45 cap, and flow controller. Solvent reservoirs and GL-45 connectors are sold separately.
TRUSTED_SUSTAINABILITY
Specifications
Column Type Flash Chromatography
For Use With (Application) Flash Chromatography
Capacity (Metric) 2000 mL
Product Line EZSafe
EZ Safe is a registered trademark of Gerresheimer Glass, Inc.