Learn More
MilliporeSigma™ Supelco™ EZSafe™ Flash-Chromatography Column with Reservoir and Fritted Disc
Catalog No. 111008888 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Capacity (Metric):
600 mL
Catalog No. Capacity (Metric)
11-100-8888 600 mL
1 options
Catalog No. 11-100-8888 Supplier MilliporeSigma™ Supelco™ Supplier No. Z4159871EA
Add to Cart
Add to Cart
Flash chromatography is useful for rapid, preparative separations with moderate resolutions.
Low pressure (max. 7 psig) medium wall columns with reservoir. Greaseless PTFE insert with GL-45 screw-thread attaches flow control valve to column. Precision all-PTFE valve and drip-tip.
TRUSTED_SUSTAINABILITY
Specifications
Column Type Flash Chromatography
For Use With (Application) Flash Chromatography
Capacity (Metric) 600 mL
Product Line EZSafe
EZ Safe is a registered trademark of Gerresheimer Glass, Inc.