Learn More
Alkali Scientific EZseal™ StripFoil™ for 8-Strip PCR Tubes, 8 Strips per Sheet, 400 Sheets
SDP
Catalog No. 502388679
Encompass_Preferred
EZseal™ StripFoil™ For sealing a 8 strip PCR Tubes or a single row of a 96-well plate. An economical and more user-friendly alternative to PCR strip-caps; no sore thumbs guaranteed! Precision cut to a smaller dimension for sealing one PCR 8-tube strip or a single row of a 96-well plate whenever rows must be selectively protected or accessed. FilmStrips remove easily with no splash back as can occur with strip-caps. Certified DNase- & RNase- free. Features an extra strong, thick adhesive to minimize evaporation. Each sheet contains 8 strips on a continuous liner. A non-locking workstation is recommended for sample prep (e.g. 96-well round-boom plate).
Catalog No. 50-238-8679 Supplier Alkali Scientific Supplier No. SF13ML8
EZseal™ StripFoil™ For sealing a 8 strip PCR Tubes or a single row of a 96-well plate. An economical and more user-friendly alternative to PCR strip-caps; no sore thumbs guaranteed! Precision cut to a smaller dimension for sealing one PCR 8-tube strip or a single row of a 96-well plate whenever rows must be selectively protected or accessed. FilmStrips remove easily with no splash back as can occur with strip-caps. Certified DNase- & RNase- free. Features an extra strong, thick adhesive to minimize evaporation. Each sheet contains 8 strips on a continuous liner. A non-locking workstation is recommended for sample prep (e.g. 96-well round-boom plate).