Learn More
STEMCELL Technologies EasySep™ Human Whole Blood and Bone Marrow CD138 Positive Selection Kit II, For processing: 60 mL whole blood or bone marrow processed
SDP
Catalog No. NC1487894
Encompass
Immunomagnetic positive selection of human CD138+ cells from whole blood and bone marrow (e.g. normal or malignant plasma cells for multiple myeloma research)
This item is not returnable. View return policy
Catalog No. NC1487894 Supplier STEMCELL Technologies Supplier No. 17887
Immunomagnetic positive selection of human CD138+ cells from whole blood and bone marrow (e.g. normal or malignant plasma cells for multiple myeloma research)