Learn More
Amsbio LLC "EZSPHERE 24-well Plate [Diameter: 500um, Depth: 100um, No. of Well: 470/well]"
Catalog No. 502566213
Encompass
EZSPHERE 24-well Plate [Diameter: 500um Depth: 100um No. of Well: 470/well]
Catalog No. 50-256-6213 Supplier Amsbio LLC Supplier No. AMS.4820900SP
Add to Cart
Add to Cart
EZSPHERE 24-well Plate [Diameter: 500um Depth: 100um No. of Well: 470/well]