Learn More
Amsbio LLC EZSPHERE 96-well Plate [Diameter: 500um, Depth: 100um, No. of Well: 95/well]
Catalog No. 502566212
Encompass
EZSPHERE 96-well Plate [Diameter: 500um Depth: 100um No. of Well: 95/well]
Catalog No. 50-256-6212 Supplier Amsbio LLC Supplier No. AMS.4860900SP
Add to Cart
Add to Cart
EZSPHERE 96-well Plate [Diameter: 500um Depth: 100um No. of Well: 95/well]