Learn More
MESA LABS BOZEMAN MFG FACILITY EZTest® for Steam / Flash 25/pk
SDP
Catalog No. NC1208399
Encompass
Self-contained biological indicator for monitoring steam. Contains bacterial spores on a small carrier pack­aged within a small, thermoplastic culture tube. Inside the thermoplastic culture tube is a sealed-glass ampoule of specially-formulated soybean casein digest culture medium containing a color indicator which turns a dramatic yellow when spores grow. Easy-to-use and produces visual results within 24 hours
Catalog No. NC1208399 Supplier MESA LABS BOZEMAN MFG FACILITY Supplier No. EZS/625
Self-contained biological indicator for monitoring steam. Contains bacterial spores on a small carrier pack­aged within a small, thermoplastic culture tube. Inside the thermoplastic culture tube is a sealed-glass ampoule of specially-formulated soybean casein digest culture medium containing a color indicator which turns a dramatic yellow when spores grow. Easy-to-use and produces visual results within 24 hours