Learn More
Foxx Life Sciences EZBiopure Titanium Round Bottle Cap Assembly, MTO, 250mL, Single Use Technology with Flexelene Tubing, Plug and GL45 VersaCap, 20/CS, Sterile
Catalog No. 502979011
Encompass_Preferred
Foxx Life Sciences presents its Single-Use Sterile EZBiopure Titanium Round Media Bottle Cap Assembly with tubing and vent filters. These assemblies let customers choose fittings (CPC MPC/AQS, plug, or luer) and TPE brand tubing (EZBio, C-Flex, AdvantaFlex, EZBioWeld, Flexlene, or PureWeld).
Utilizing Foxx's patented VersaCap technology, no machining is required, making them ideal for single-use applications.
Each EZBiopure Titanium Round (TR) MTO cap assembly includes a GL45 open cap with adapter, GL45 close cap for storage, TPE tubing (ID 1/8 in, OD 1/4 in), fittings, a dip tube for precise draw, a 0.22 µm vent filter, and a ratchet-style pinch clamp. Each unit is double-bagged and gamma sterilized in the U.S.A.
Features
Ideal for single-use applications, reducing reliance on costly stainless-steel systems
Made entirely with USP Class VI components
Gamma compatible, meeting a sterile assurance level (SAL) of 10⁻⁶
Catalog No. 50-297-9011 Supplier Foxx Life Sciences Supplier No. 5205440FLS
Foxx Life Sciences presents its Single-Use Sterile EZBiopure Titanium Round Media Bottle Cap Assembly with tubing and vent filters. These assemblies let customers choose fittings (CPC MPC/AQS, plug, or luer) and TPE brand tubing (EZBio, C-Flex, AdvantaFlex, EZBioWeld, Flexlene, or PureWeld).
Utilizing Foxx's patented VersaCap technology, no machining is required, making them ideal for single-use applications.
Each EZBiopure Titanium Round (TR) MTO cap assembly includes a GL45 open cap with adapter, GL45 close cap for storage, TPE tubing (ID 1/8 in, OD 1/4 in), fittings, a dip tube for precise draw, a 0.22 µm vent filter, and a ratchet-style pinch clamp. Each unit is double-bagged and gamma sterilized in the U.S.A.
Features
Ideal for single-use applications, reducing reliance on costly stainless-steel systems
Made entirely with USP Class VI components
Gamma compatible, meeting a sterile assurance level (SAL) of 10⁻⁶