Learn More
Medchemexpress LLC Ezurpimtrostat hydrochloride | 1914148-73-4 | 99.3% | 459.45 | 10 MM * 1 ML
SDP
Catalog No. 5000347907
Encompass_Preferred
Ezurpimtrostat hydrochloride (compound 2-3) is a potent and orally active anti-fibrotic agent. It significantly reduces liver fibrosis in DEN (diethyl nitrosamine) cirrhotic rat models, and can be utilized for research related to fibrosis, cancer, autophagy, and diseases involving cathepsins B (CTSB), L (CTSL), and D (CTSD).
  • Potent and orally active anti-fibrotic agent.
  • Significantly reduces liver fibrosis in DEN cirrhotic rat models.
  • Applicable for research in fibrosis, cancer, autophagy, and cathepsins B, L, and D related diseases.
  • Purity of 99.29%.
Catalog No. 50-003-47907 Supplier Medchemexpress LLC Supplier No. HY137978A10MM/1ML
Ezurpimtrostat hydrochloride (compound 2-3) is a potent and orally active anti-fibrotic agent. It significantly reduces liver fibrosis in DEN (diethyl nitrosamine) cirrhotic rat models, and can be utilized for research related to fibrosis, cancer, autophagy, and diseases involving cathepsins B (CTSB), L (CTSL), and D (CTSD).
  • Potent and orally active anti-fibrotic agent.
  • Significantly reduces liver fibrosis in DEN cirrhotic rat models.
  • Applicable for research in fibrosis, cancer, autophagy, and cathepsins B, L, and D related diseases.
  • Purity of 99.29%.