Learn More
TARGETMOL CHEMICALS INC Ezurpimtrostat hydrochloride
SDP
Catalog No. 502879869
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Ezurpimtrostat hydrochloride (GNS561 hydrochloride) is an orally available palmitoyl protein thioesterase-1 inhibitor with antifibrotic, antiviral, and anticancer effects that blocks cancer cell proliferation by interacting with palmitoyl protein thioesterase to inhibit late autophagy and dose-dependently increase lysosomal accumulation. Purity 99.84%
Catalog No. 50-287-9869 Supplier TARGETMOL CHEMICALS INC Supplier No. T628821MG
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Ezurpimtrostat hydrochloride (GNS561 hydrochloride) is an orally available palmitoyl protein thioesterase-1 inhibitor with antifibrotic, antiviral, and anticancer effects that blocks cancer cell proliferation by interacting with palmitoyl protein thioesterase to inhibit late autophagy and dose-dependently increase lysosomal accumulation. Purity 99.84%