Learn More
Sigma Aldrich Fine Chemicals Biosciences EZviewTM Red Anti c Myc Af
Catalog No. 502914711
Encompass
EZ view Red Anti--c-Myc Affinity Gel is a resin that consists of the Anti-c-Myc rabbit antibody covalently bonded to 4% Agarose beads. The affinity gel is used to bind c-Myc fusion proteins to samples such as cell lysates and tissue for purification of c-Myc-tagged proteins in preparation of immunoprecipitation assays. Red dye enhances visability for more efficient results.Agarose beads bind at N-terminal Met-N-terminal and C-terminal c-Myc fusion proteins from E. coli and mammalian cells.
Catalog No. 50-291-4711 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. E66545X1ML
EZ view Red Anti--c-Myc Affinity Gel is a resin that consists of the Anti-c-Myc rabbit antibody covalently bonded to 4% Agarose beads. The affinity gel is used to bind c-Myc fusion proteins to samples such as cell lysates and tissue for purification of c-Myc-tagged proteins in preparation of immunoprecipitation assays. Red dye enhances visability for more efficient results.Agarose beads bind at N-terminal Met-N-terminal and C-terminal c-Myc fusion proteins from E. coli and mammalian cells.