Learn More
Sigma Aldrich Fine Chemicals Biosciences EZviewTM Red Protein A Aff
Supplier:  Sigma Aldrich Fine Chemicals Biosciences P64865X1ML
Encompass
EZview(TM) Red Protein A Affinity Gel is a highly visible red colored Protein A affinity gel. The affinity resin contains Staphylococcus aureus Protein A covalently attached to crosslinked 4% agarose beads. The affinity gel is used to bind the fC portion of the IgG of an antibody to samples such as cell lysates and tissue. The immune complexes bound to the EZview(TM) Red Protein A Affinity Gel are recovered by centrifugation. Red dye enhances visibility for more efficient results.
Catalog No. 50-291-4340