Learn More
Sigma Aldrich Fine Chemicals Biosciences EZviewTM Red Protein G Aff
Supplier:  Sigma Aldrich Fine Chemicals Biosciences E34031ML
Encompass
EZ view Red Protein G Affinity Gel is Protein G covalently bonded to 4% Agarose beads. The affinity gel is used to bind the fC portion of the IgG of an antibody to samples such as cell lysates and tissue for purification of proteins in preparation of immunoprecipitation assays. Red dye enhances visability for more efficient results.
Catalog No. 50-290-4551