Learn More
Bellco Glass, Inc. EZwaste 8" Safety Funnel, HDPE, Red Lid, VersaCap 38-430mm, 1/EA
SDP
Supplier:  Bellco Glass, Inc. 452800145
Encompass_Preferred
EZwaste 8″ Safety Funnel features a latching lid with gasket to create an air-tight seal and to keep VOCs from evaporating into the air. The EZwaste Safety Funnel is the ultimate lab safety product for liquid waste collection. Our safety funnels can be used as both a sealed waste container and a solvent funnel. Our Safety funnel also has a built-in interior filter to reduce the loss of stir sticks or other particles in the waste liquid. Applications include but not limited Pharmaceutical, biotech, and Laboratories.Prevents 99.9% of VOCs emission.11″ stem reduces the emission of VOCs in the labMade from HDPE MaterialMeets most local, state and federal safety regulations and requirements.Reusable and resists nearly all corrosive chemicals. Material: HDPECapacity: 2LCap Size: 38-430Dimensions: 11″ Length x 9″ widthStem Length: 11″
Catalog No. 50-255-0617