Learn More
Bellco Glass, Inc. EZwaste, Filter, Exhaust, Large, 1/4" NPT Female Port, HDPE, 1/EA
SDP
Supplier:  Bellco Glass, Inc. 452800133
Encompass_Preferred
EZwaste XL Solvent Waste filter replacements come in a pack of 1.The Replacement Chemical Exhaust Filter EZwaste XL Solvent Waste System is designed to be an affordable solution for HPLC liquid waste disposal. Each system is manufactured to safely collect waste from two HPLC machines. This system features an activated carbon filter which traps volatile organic compounds from being released into the air. This prevents chemicals from being inhaled by lab workers and provides an easy mechanism of disposal for vapor wastes. The EZwaste XL Solvent Waste system also features a closed loop design which protects against leaks and spills, improving overall lab safety.* Depending on application, carbon exhaust filter life cycle may vary Height: 10.5″ (266.7 mm)OD: 2.68″ (68.0 mm)Filter thread: 1/4″
Catalog No. 50-255-0629