Learn More
Foxx Life Sciences EZwaste HPLC Solvent Waste System, Replacement Fittings, 1/8 Inch OD Tube Fittings, Blue Nut with Yellow Ferrule, 1/4-28 Thread, 6/PK
Catalog No. 501178770
Encompass_Preferred
330-0931-OEM: EZwaste Replacement Fittings, 1/8" OD, 6/pk. Foxx Life Sciences' replacement fittings secure waste tubes to carbos, providing the closed system necessary to prevent spills. The EZwaste Solvent Waste System is designed to be an affordable solution for HPLC liquid waste disposal. Each system is manufactured to safely collect waste from two HPLC machines. This system features an activated carbon filter, which traps volatile organic compounds from being released into the air. This prevents chemicals from being inhaled by lab workers, and provides an easy mechanism of disposal for vapor wastes. The EZwaste Solvent Waste system also features a closed-loop design which protects against leaks and spills, improving overall lab safety.
Catalog No. 50-117-8770 Supplier Foxx Life Sciences Supplier No. 3300931OEM
330-0931-OEM: EZwaste Replacement Fittings, 1/8" OD, 6/pk. Foxx Life Sciences' replacement fittings secure waste tubes to carbos, providing the closed system necessary to prevent spills. The EZwaste Solvent Waste System is designed to be an affordable solution for HPLC liquid waste disposal. Each system is manufactured to safely collect waste from two HPLC machines. This system features an activated carbon filter, which traps volatile organic compounds from being released into the air. This prevents chemicals from being inhaled by lab workers, and provides an easy mechanism of disposal for vapor wastes. The EZwaste Solvent Waste system also features a closed-loop design which protects against leaks and spills, improving overall lab safety.