Learn More
Bellco Glass, Inc. EZWaste HPLC VersaCap 53mm Solvent Waste Cap Assembly W/ Exhaust Filter, 6 X Ports OD Tube-3.2 mm (1/8") & 1.6 mm (1/16"), 1/EA
SDP
Supplier:  Bellco Glass, Inc. 452800113
Encompass_Preferred
EZWaste HPLC VersaCap 53mm Solvent Waste Cap Assembly W/ Exhaust Filter, 6 X Ports OD Tube-3.2 mm (1/8") & 1.6 mm (1/16"), 1/EA
Catalog No. 50-255-0649