Learn More
Foxx Life Sciences EZwaste HPLC Solvent Waste System, Breakthrough Indicators for Detecting Carbon Filter Saturation, 5/PK
Catalog No. 501178703
Encompass_Preferred
330-0908-OEM: EZwaste Breakthrough Indicator (5pk). EZwaste Replacement Breakthrough Indicators come in packs of 1, 2, 5, 10, 25, or 50. Foxx Life SciencesBreakthrough Indicator is a qualitative colorimetric indicator for the exhaustion and end-of-service life of low-flow filters. The indicator changes from orange to red when the filter needs to be replaced. The indicator is designed to provide real-time indication of the breakthrough of HPLC solvents vapors, including: Acetone Acetonitrile Chloroform DCM/Methylene Chloride (Dichloromethane) Ethanol Ethyl Acetate HFIP Hexafluoroisopropanol Methanol Phenol Sulfolane THF (Tetrahydrofuran)
Catalog No. 50-117-8703 Supplier Foxx Life Sciences Supplier No. 3300908OEM
330-0908-OEM: EZwaste Breakthrough Indicator (5pk). EZwaste Replacement Breakthrough Indicators come in packs of 1, 2, 5, 10, 25, or 50. Foxx Life SciencesBreakthrough Indicator is a qualitative colorimetric indicator for the exhaustion and end-of-service life of low-flow filters. The indicator changes from orange to red when the filter needs to be replaced. The indicator is designed to provide real-time indication of the breakthrough of HPLC solvents vapors, including: Acetone Acetonitrile Chloroform DCM/Methylene Chloride (Dichloromethane) Ethanol Ethyl Acetate HFIP Hexafluoroisopropanol Methanol Phenol Sulfolane THF (Tetrahydrofuran)