Learn More
Bellco Glass, Inc. EZwaste Replacement Fittings, 1/2-20 Plugs, 20/PK
SDP
Supplier:  Bellco Glass, Inc. 452800157
Encompass_Preferred
The 1/2-20″ Plugs Pack is the replacement fittings for EZwaste Solvent Waste System’s fittings. It is used to cover any unused 1/4″ ports. It includes (20) 1/2-20″ plugs. These plugs provide closed system necessary to prevent spills.The EZwaste Solvent Waste System is designed to be an affordable solution for HPLC liquid waste disposal. Each system is manufactured to safely collect waste from two HPLC machines. This system features an activated carbon filter which traps volatile organic compounds from being released into the air. This prevents chemicals from being inhaled by lab workers and provides an easy mechanism of disposal for vapor wastes. The EZwaste Solvent Waste system also features a closed loop design which protects against leaks and spills, improving overall lab safety.* Depending on application, carbon exhaust filter life cycle may vary Material: HDPEColor: Yellow
Catalog No. 50-255-0605